1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
Thermo Scientific Chemicals

1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%

CAS: 827614-66-4 | C13H23BN2O2 | 250.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 439730010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS827614-66-4
IUPAC Name1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular FormulaC13H23BN2O2
InChI KeyYMEBZRNYQBODKB-UHFFFAOYSA-N
SMILESCC(C)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only