(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%
(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%
(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%
Thermo Scientific Chemicals

(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%

CAS: 156311-83-0 | C17H27F6N7OP2 | 521.39 g/mol
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 440610010
Price (JPY)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Assay Percent Range98%
CAS Max %100.0
CAS Min %97.5
CAS156311-83-0
Chemical Name or Material(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only