PrepSEQ™ Rapid Spin Sample Preparation Kit - Extra Clean
The PrepSEQ™ Rapid Spin Sample Preparation Kit – Extra Clean is recommended for foods with a high lipid content, suchRead more
Have Questions?
Catalog Number
Quantity
4413269
1 kit
Catalog number 4413269
Price (USD)
487.00
Each
Quantity:
1 kit
Price (USD)
487.00
Each
The PrepSEQ™ Rapid Spin Sample Preparation Kit – Extra Clean is recommended for foods with a high lipid content, such as infant formula and chicken wing samples. The kit was developed for the rapid preparation of DNA from food and environmental samples for use in PCR amplification reactions. Using a simple spin protocol, PrepSEQ™ Rapid Spin efficiently inactivates PCR inhibitors. For laboratories involved in food and environmental testing, PCR-based methods are rapidly becoming the standard for accurate, sensitive bacterial detection. This has created the demand for a sample preparation method that is fast, cost-effective and capable of extracting high-quality DNA from a broad range of sample types. While such methods have been developed for dedicated, high-throughput laboratories, there is a growing need for methods suitable to low-to-medium throughput applications. The PrepSEQ™ Rapid Spin Kit – Extra Clean meets this need.
• Designed for Food Safety and Environmental Monitoring applications • Supports low-to-medium throughput work loads (up to 100 samples a day) • Inactivates PCR inhibitors so you will most likely not have to repeat the template preparation step • Uses a simple spin protocol Optimized for PCR-based Food Safety and Environmental Monitoring For laboratories involved in food safety and environmental monitoring, PCR-based methods are rapidly becoming the standard for accurate, sensitive bacterial detection. Use this sample preparation method for fast, cost-effective extraction of high-quality DNA from a broad range of sample types including chocolate, soft cheeses, and soil.
Designed for Laboratories that Prepare up to 100 Samples a Day Kits exist for dedicated, high-throughput laboratories, but now you have a kit that is suitable for low-to-medium throughput applications. This kit is designed for laboratories that routinely prepare up to 100 samples a day.
Removes PCR Inhibitors The PrepSEQ™ Rapid Spin Sample Preparation Kit offers a simple, cost-effective way to prepare bacterial DNA from broth cultures. During the extraction process, the media inside the spin column removes PCR inhibitors from the sample, allowing the bacteria to pass through into the sample collection tube where it can be stored or used for PCR.
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Final Product TypeBacterial DNA
For Use With (Application)Real-Time Quantitative PCR (qPCR), PCR