Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000
Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000
Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000
Thermo Scientific Chemicals

Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000

Dextran sulfate sodium salt, C9H11NO2, CAS Number-9011-18-1, americaine, anaesthesin, anesthesin, benzocaine, ethoform, ethyl aminobenzoate, ethyl p-aminobenzoate, norcaine, orthesin, parathesin, 25g, CHEBI:116735, BLFLLBZGZJTVJG-UHFFFAOYSA-N, ethyl 4-aminobenzoate, MFCD00081551, 165.192 | CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 441490250
Price (USD)
226.65
Special offer
Online exclusive
Ends: 30-Jun-2026
267.00
Save 40.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
226.65
Special offer
Online exclusive
Ends: 30-Jun-2026
267.00
Save 40.35 (15%)
Each
Chemical Identifiers
CAS9011-18-1
IUPAC NameDextran sulfate sodium salt
MDL NumberMFCD00081551
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+75° to +105° (20°C, 589 nm) (6% in water)
Sulfur (S)16 to 20 %
Specific viscosity0.018 to 0.032 (1.0M in NaCl, 25°C)
Molecular weight6500 to 10000
Appearance (Color)White to light yellow to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only