4-Chloro-3-fluoropyridine, 95%
4-Chloro-3-fluoropyridine, 95%
4-Chloro-3-fluoropyridine, 95%
Thermo Scientific Chemicals

4-Chloro-3-fluoropyridine, 95%

CAS: 2546-56-7 | C5H3ClFN | 131.53 g/mol
Quantity:
5 g
25 g
Catalog number 443160050
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2546-56-7
IUPAC Name4-chloro-3-fluoropyridine
Molecular FormulaC5H3ClFN
InChI KeyBEQUUSCRAKEKQM-UHFFFAOYSA-N
SMILESFC1=C(Cl)C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94.0 %
Appearance (Color)Colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only