Trichloroacetic anhydride, 95%
Trichloroacetic anhydride, 95%
Trichloroacetic anhydride, 95%
Thermo Scientific Chemicals

Trichloroacetic anhydride, 95%

CAS: 4124-31-6 | C4Cl6O3 | 308.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 446091000
Price (USD)
391.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
391.00
Each
Chemical Identifiers
CAS4124-31-6
IUPAC Name2,2,2-trichloroacetyl 2,2,2-trichloroacetate
Molecular FormulaC4Cl6O3
InChI KeyMEFKFJOEVLUFAY-UHFFFAOYSA-N
SMILESClC(Cl)(Cl)C(=O)OC(=O)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4830 to 1.4870 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only