BOC-5-Aminopentanoic acid, 97%
BOC-5-Aminopentanoic acid, 97%
BOC-5-Aminopentanoic acid, 97%
Thermo Scientific Chemicals

BOC-5-Aminopentanoic acid, 97%

CAS: 27219-07-4 | C10H19NO4 | 217.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 448240050
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS27219-07-4
IUPAC Name5-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Molecular FormulaC10H19NO4
InChI KeyGFMRZAMDGJIWRB-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point48°C to 54°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only