N-BOC-4-oxo-L-proline, 98%
N-BOC-4-oxo-L-proline, 98%
N-BOC-4-oxo-L-proline, 98%
Thermo Scientific Chemicals

N-BOC-4-oxo-L-proline, 98%

CAS: 84348-37-8 | C10H15NO5 | 229.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4492800101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 449280010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS84348-37-8
IUPAC Name1-[(tert-butoxy)carbonyl]-4-oxopyrrolidine-2-carboxylic acid
Molecular FormulaC10H15NO5
InChI KeyCKYGSXRXTIKGAJ-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CC(=O)CC1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Form)Crystalline solid
Melting point157°C to 163°C
Specific optical rotation+18° to +23° (20°C, 589 nm) (c=0.5, acetone)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only