Choline bitartrate, 97%
Choline bitartrate, 97%
Choline bitartrate, 97%
Thermo Scientific Chemicals

Choline bitartrate, 97%

CAS: 87-67-2 | C5H14NO·C4H5O6 | 253.25 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
產品號碼 450225000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS87-67-2
IUPAC Name(2-hydroxyethyl)trimethylazanium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
Molecular FormulaC9H19NO7
InChI KeyQWJSAWXRUVVRLH-LREBCSMRSA-M
SMILESC[N+](C)(C)CCO.O[C@H]([C@@H](O)C([O-])=O)C(O)=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White
Infrared spectrumConforms
Water=<0.5 %
Appearance (Form)Crystalline powder
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only