Metaphosphoric acid, ACS reagent, stabilized
Metaphosphoric acid, ACS reagent, stabilized
Metaphosphoric acid, ACS reagent, stabilized
Thermo Scientific Chemicals

Metaphosphoric acid, ACS reagent, stabilized

CAS: 37267-86-0 | HO3P | 79.979 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Catalog number 450251000
Price (USD)
126.00
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
126.00
Each
Chemical Identifiers
CAS37267-86-0
IUPAC Nametrihydroxy-1,3,5,2λ⁵,4λ⁵,6λ⁵-trioxatriphosphinane-2,4,6-trione
Molecular FormulaH3O9P3
InChI KeyAZSFNUJOCKMOGB-UHFFFAOYSA-N
SMILESOP1(=O)OP(O)(=O)OP(O)(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH33.5 to 36.5 % (HPO3)
Nitrate (NO3)=<0.001 %
Arsenic (As)=<1 ppm
KMnO4-reducing substance=<0.02 % (as H3PO3)
Appearance (Color)Colorless to white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only