5-Amino-1-methyl-1H-pyrazole, 97%
5-Amino-1-methyl-1H-pyrazole, 97%
5-Amino-1-methyl-1H-pyrazole, 97%
Thermo Scientific Chemicals

5-Amino-1-methyl-1H-pyrazole, 97%

CAS: 1192-21-8 | C4H7N3 | 97.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 452610250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1192-21-8
IUPAC Name1-methyl-1H-pyrazol-5-amine
Molecular FormulaC4H7N3
InChI KeyJESRNIJXVIFVOV-UHFFFAOYSA-N
SMILESCN1N=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
Melting point69°C to 77°C
Appearance (Color)White to yellow
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only