Injection Port Liners, Split/Splitless Mixed Liner Sample Pack
Injection Port Liners, Split/Splitless Mixed Liner Sample Pack
Injection Port Liners, Split/Splitless Mixed Liner Sample Pack
Injection Port Liners, Split/Splitless Mixed Liner Sample Pack
Thermo Scientific™
Injection Port Liners, Split/Splitless Mixed Liner Sample Pack
Use these injection port liners to optimize performance of Agilent GC instruments.
Catalog NumberQuantity
453AG0015
Catalog number 453AG001
Price (JPY)
-
Quantity:
5
Optimize the performance of Agilent™ GC systems with our specifically designed Thermo Scientific™ Injection Port Liners for Agilent instruments. Highly deactivated and produced to exacting tolerances, these injection liners ensure exceptional reproducibility.
  • Highly deactivated borosilicate glass
  • Quartz wool packing material
  • ID × OD × L: 4 × 6.3 × 78.5mm
Specifications
Description453AG001
For Use With (Application)Split/Splitless
For Use With (Equipment)Thermo Scientific TRACE 1300/1600 series and Agilent GC instruments
Quantity5
Length (Metric)Mixed
Packing MaterialMixed
Product TypeSplit/Splitless Mixed Liner Sample Pack
Unit SizeEach