Acetic acid, 50%, solution in water
Acetic acid, 50%, solution in water
Acetic acid, 50%, solution in water
Acetic acid, 50%, solution in water
Thermo Scientific Chemicals

Acetic acid, 50%, solution in water

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
25 L
Catalog number 456870050
Price (USD)
-
Quantity:
5 L
Request bulk or custom format
Chemical Identifiers
CAS64-19-7
IUPAC Nameacetic acid
Molecular FormulaC2H4O2
InChI KeyQTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILESCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Titration with NaOH49.0 to 51.0 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only