Decyl decanoate, 98%
Decyl decanoate, 98%
Decyl decanoate, 98%
Thermo Scientific Chemicals

Decyl decanoate, 98%

CAS: 1654-86-0 | C20H40O2 | 312.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
Catalog number 457115000
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS1654-86-0
IUPAC Namedecyl decanoate
Molecular FormulaC20H40O2
InChI KeyXAKXZZPEUKNHMA-UHFFFAOYSA-N
SMILESCCCCCCCCCCOC(=O)CCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.4410 to 1.4430 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only