CD25 Monoclonal Antibody (OX39), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD25 Monoclonal Antibody (OX39), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (OX39), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0390-82, 46-0390-80)