BCL6 Monoclonal Antibody (BCL-DWN), PerCP-eFluor™ 710, eBioscience™ - FAQs
BCL6 Monoclonal Antibody (BCL-DWN), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for BCL6 Monoclonal Antibody (BCL-DWN), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5453-80, 46-5453-82)