Efaproxiral sodium, 98%
Efaproxiral sodium, 98%
Efaproxiral sodium, 98%
Thermo Scientific Chemicals

Efaproxiral sodium, 98%

CAS: 170787-99-2 | C20H22NNaO4 | 363.38 g/mol
製品番号(カタログ番号) 460100010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS170787-99-2
IUPAC Namesodium 2-(4-{[(3,5-dimethylphenyl)carbamoyl]methyl}phenoxy)-2-methylpropanoate
Molecular FormulaC20H22NNaO4
InChI KeySWDPIHPGORBMFR-UHFFFAOYSA-M
SMILES[Na+].CC1=CC(NC(=O)CC2=CC=C(OC(C)(C)C([O-])=O)C=C2)=CC(C)=C1
さらに表示
Loss on drying=<13 %
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only