Ammonium chloride, for molecular biology
Thermo Scientific Chemicals

Ammonium chloride, for molecular biology

CAS: 12125-02-9 | ClH4N | 53.49 g/mol
Catalog NumberQuantity
464595000500 g
Catalog number 464595000
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS12125-02-9
IUPAC Nameamine hydrochloride
Molecular FormulaClH4N
InChI KeyNLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILESN.Cl
View more
SpecificationsSpecification SheetSpecification Sheet
pH4.5 to 5.5 (5 % in water)
Phosphate (PO4)=<2 ppm
RNase activityNone detected
Appearance (Form)Crystalline powder or crystals and/or chunks
Sulfate (SO4)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only