Simethicone
Thermo Scientific Chemicals

Simethicone

CAS: 8050-81-5 | C6H18O4Si3 | 238.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 465170500
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS8050-81-5
IUPAC Name3,3,5,5-tetramethyl-2,4-dioxa-3,5-disilahexane; silanedione
Molecular FormulaC6H18O4Si3
InChI KeyAMTWCFIAVKBGOD-UHFFFAOYSA-N
SMILESO=[Si]=O.CO[Si](C)(C)O[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to grey
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Refractive index1.4000 to 1.4100 (20°C, 589 nm)
Viscosity475 to 525 CP (25°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only