Product Image
Thermo Scientific Chemicals

Lacosamide

CAS: 175481-36-4 | C13H18N2O3 | 250.30 g/mol
Quantity:
100 mg
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465321000
Price (JPY)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS175481-36-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Water=<0.5 %
Specific optical rotation+14° to +18° (20°C, 589 nm) (0.1g in 10 ml CH3OH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only