(-)-Arctigenin
Thermo Scientific Chemicals

(-)-Arctigenin

Quantity:
250 mg
1 g
Catalog number 466040010
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7770-78-7
IUPAC Name(3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[(4-hydroxy-3-methoxyphenyl)methyl]oxolan-2-one
Molecular FormulaC21H24O6
InChI KeyNQWVSMVXKMHKTF-JKSUJKDBSA-N
SMILESCOC1=CC=C(C[C@H]2COC(=O)[C@@H]2CC2=CC=C(O)C(OC)=C2)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Melting point98°C to 102°C
Water=<3 %
HPLC>=97.5 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only