Potassium phosphate tribasic, 90+%, pure
Thermo Scientific Chemicals

Potassium phosphate tribasic, 90+%, pure

CAS: 7778-53-2 | K3O4P | 212.27 g/mol
Quantity:
1 kg
5 kg
25 kg
Catalog number 467330010
Price (JPY)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Assay Percent Range90+%
CAS7778-53-2
Chemical Name or MaterialPotassium phosphate tribasic
EINECS Number231-907-1
Formula Weight212.27
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only