CD45.2 Monoclonal Antibody (104), eFluor™ 450, eBioscience™ - FAQs
CD45.2 Monoclonal Antibody (104), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD45.2 Monoclonal Antibody (104), eFluor™ 450, eBioscience™ - FAQs (48-0454-82, 48-0454-80)