Barcoded Latch Racks for Matrix™ Storage Tubes
Thermo Scientific™

Barcoded Latch Racks for Matrix™ Storage Tubes

Create custom arrays with empty ANSI-format racks featuring 2D and linear barcodes and human readable type on three sides and a 2D barcode on the base of the rack.
Catalog NumberFor Use With (Equipment)No. per Case
4898-BR500μL Tubes10
4897-BR1.0mL Screwtop Tubes5
4900-BR500μL Screwtop Tubes5
Catalog number 4898-BR
Price (JPY)
18,820
Case of 10
For Use With (Equipment):
500μL Tubes
No. per Case:
10
Enhance sample storage at low temperatures, including vapor phase Liquid Nitrogen, with Thermo Scientific™ Matrix™ barcoded latch racks. Barcoded Latch Racks feature a linear barcode, 2D barcode and human readable rack identification on three sides of the rack for the ultimate in sample tracking. Use in combination with Thermo Scientific 2D barcoded tubes, barcode readers, arrayers and capping equipment.

Superior Storage Rack Design

  • Storage in robot-friendly ANSI-footprint rack
  • Racks feature linear barcode, human readable ID and 2D barcode for ultimate sample tracking regardless of your barcode system
  • Rack barcodes are compatible with our range of VisionMate readers
  • Hinged lid rests in open position on rack while conducting manual pipetting to prevent contamination of lid sitting on bench

Latch Racks Save Precious Space

  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Latch rack lid can be lifted robotically for tube access with high-throughput automated applications
Specifications
Barcode2D Datamatrix; Linear EAN Code 128; Human Readable
Capacity (English)0.0169 fl oz.
ColorWhite
DescriptionEmpty Latch Rack
For Use With (Equipment)500μL Tubes
Holds2D Matrix tubes
No. per Case10
Packaging Quantity10 Case
Product LineMatrix
Capacity (Metric)0.5 mL
MaterialPlastic
Unit SizeCase of 10