Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL-13 Antibodies

          Invitrogen

          IL-13 Monoclonal Antibody (eBio13A), Alexa Fluor™ 488, eBioscience™

          Advanced Verification
          42 Published Figures
          56 References
          View all (89) IL-13 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite IL-13 Monoclonal Antibody (eBio13A), Alexa Fluor™ 488, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (42)
          • Advanced Verification (1)
          IL-13 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          IL-13 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 44

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-13 Antibody (53-7133-80) in Flow

          Intracellular staining of Th2-polarized and restimulated CD4+ splenocytes with Anti-Mouse CD4 PerCP-eFluor® 710 (Product # 46-0042-82) and 0.125 µg of Rat IgG1 K Isotype Control Alexa Fluor® 488 (Product # 53-4301-80) (left) or 0.125 µg of Anti-Mouse IL-13 Alexa Fluor® 488 (right). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry, ELISA (Flow, ELISA)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Flow Cytometry (Flow)
          IL-13 Antibody in Neutralization (Neu)
          IL-13 Antibody

          Product Details

          53-7133-80

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 52 publications 52 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Neutralization (Neu)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG1, kappa

          Recommended Isotype Control

          Rat IgG1 kappa Isotype Control (eBRG1), Alexa Fluor™ 488, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBio13A

          Conjugate

          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          • Unconjugated
          • APC-eFluor 780
          • Brilliant UV 737
          • Brilliant UV 805
          • eFluor 450
          • eFluor 660
          • Functional Grade
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Request custom conjugation

          Excitation/Emission Max

          499/520 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2016639

          Product Specific Information

          Description: The eBio13A antibody reacts with mouse IL-13. IL-13 is a cytokine produced mainly by Th2 cells, but also by antigen-primed CD8 T cells. IL-13 has a strong involvement in allergic inflammation and parasitic clearing and in cancer models has been shown to have either inhibitory or stimulatory activity depending on the tumor. In humans, IL-13 is found to play a role in isotype switching in B cells. IL-13 is implicating in down modulating macrophage activity, through the reduction of pro-inflammatory cytokines (IL-1, IL-6, IL-8, IL-10, IL-12)

          Applications Reported: This eBio13A antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

          Applications Tested: This eBio13A antibody has been tested by intracellular staining and flow cytometric analysis of Th2 polarized CD4 cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: ALRH; ALRH antibody; BHR1; il 13; IL-13; ILN; Interleukin; Interleukin-13; Interleukin13; MGC116786; MGC116788; MGC116789; NC30 antibody; P600 antibody; T-cell activation protein P600

          View more View less

          Gene Aliases: Il-13; Il13

          View more View less

          UniProt ID: (Mouse) P20109

          View more View less

          Entrez Gene ID: (Mouse) 16163

          View more View less

          Function(s)
          cytokine activity cytokine receptor binding interleukin-13 receptor binding
          Process(es)
          microglial cell activation positive regulation of immunoglobulin production inflammatory response immune response regulation of proton transport positive regulation of B cell proliferation positive regulation of connective tissue growth factor production negative regulation of NAD(P)H oxidase activity response to nicotine positive regulation of tyrosine phosphorylation of Stat6 protein positive regulation of macrophage activation positive regulation of ion transport positive regulation of mast cell degranulation positive regulation of smooth muscle cell proliferation positive regulation of protein secretion positive regulation of release of sequestered calcium ion into cytosol cellular response to cytokine stimulus negative regulation of transforming growth factor beta production negative regulation of neuron death negative regulation of complement-dependent cytotoxicity positive regulation of pancreatic stellate cell proliferation negative regulation of endothelial cell apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5df784475-jh7xd:80/100.66.79.197:80.
          git-commit: 61700cebeb8889bf3cb500b1da5d059e71269fa5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline