CaptureSelect™ Human Growth Hormone (HGH) RoboColumn™ Pre-packed columns
Thermo Scientific™

CaptureSelect™ Human Growth Hormone (HGH) RoboColumn™ Pre-packed columns

Thermo Scientific™ CaptureSelect™ RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.
Have Questions?
카탈로그 번호수량
59431602001 Strip (8 Pieces), 200 μL
카탈로그 번호 5943160200
제품 가격(KRW)
885,000
Each
수량:
1 Strip (8 Pieces), 200 μL
제품 가격(KRW)
885,000
Each

CaptureSelect RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

The CaptureSelect Human Growth Hormone (hGH) Affinity Matrix in this robocolumn has been specifically designed for the purification of human growth hormone from recombinant sources. The high selectivity and yields obtained using CaptureSelect Human Growth Hormone Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step.

Features of this affinity matrix include:

  • Highly selective for recombinant hGH
  • Efficient purification of recombinant hGH with high yields and retained activity
  • Excellent scalability
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Characteristics

  • Column size: 200 μL, 1 strip, 8 columns (8-row)
  • Column dimension: 5-mm diameter, 10-mm bed height
  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect hGH affinity ligand
  • Ligand coupling: epoxide coupling
  • Binding capacity: ∼3 mg hGH/mL resin depending on flow rate, column height, and contact time
  • Elution conditions: 20 mM citric acid, pH 3.0
  • Formulation buffer: 20% (v/v) ethanol
  • Operating pressure: 2 bar (maximum)
  • Flow rate: 16–150 cm/hr

RoboColumn is a trademark of Repligen.

사양
컬럼 유형Affinity, Prepacked
형식Robo Column
수량1 Strip (8 Pieces), 200 μL
용량(미터법)200 μL
입자 크기65 ±10 μm
제품라인CaptureSelect
Unit SizeEach