Vanquish™ Pump Mixers
Actual product may vary
Vanquish™ Pump Mixers
Use these biocompatible pump mixers in Vanquish UHPLC systems for optimal mixing, including difficult agents such as TFA.
Catalog NumberDescriptionVolume (Metric)
6044.5018Mixer Set, Vanquish H Pump35 μL
6268.5120Mixer Set, Vanquish H Pump200 μL
6044.5110Static Mixer, Vanquish H/F Pumps150 μL
6044.5310Static Mixer, Vanquish H/F Pumps350 μL
6268.5310Mixer Set, Vanquish H Pump400μL
6044.3870Mixer Set, Vanquish F Pumps35 μL
6044.5100Mixer Set, Vanquish F Pumps100 μL
6044.5750AStatic Mixer, Vanquish F Pumps750 μL
6044.5450AStatic Mixer, Vanquish F Pumps1500 μL
6044.5026Capillary Mixer, Vanquish F Pumps50 μL
Catalog number 6044.5018
Price (JPY)
-
Description:
Mixer Set, Vanquish H Pump
Volume (Metric):
35 μL
Use the biocompatible Thermo Scientific™ Vanquish™ Pump Mixers with Vanquish binary, quaternary, or dual gradient UHPLC pumps to ensure the highest possible mixing efficiency for ultrahigh resolution and ultrafast separations. These mixers accomplish complete mixing of the HPLC gradient elution to achieve a smooth baseline with high signal-to-noise ratios, and accurate peak identification. Vanquish Pump Mixers are designed to reliably provide the highest possible performance over a wide application range with varying solvent ratios, solvent miscibilities, flow rates.
Specifications
DescriptionMixer Set, Vanquish H Pump
IncludesStatic mixer: 10 μL<br /> Capillary mixer: 25 μL
Volume (Metric)35 μL
For Use With (Equipment)Vanquish Binary Pump H: Default (For Use with Vanquish H Pump Only)
Product LineVanquish
TypeMixer Set
Unit SizeEach