Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • ELISA Kits ›
          • ERK1/2 ELISA Kits
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for will be discontinued once stock is depleted. Please check out our other {{productObj.targetName}} {{productObj.pendingDiscontinuationTargetLabel}} Please check out our other {{productDetailViewModel.formattedProductSubType.equals('ELISA') ? 'ELISA kits' : 'antibodies'}} or contact Technical Support with any questions.

          Invitrogen

          Multispecies ERK1/ERK2 In-Cell ELISA Kit, Near Infrared

          View all (5) ERK1/2 ELISA Kits

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Multispecies ERK1/ERK2 In-Cell ELISA Kit, Near Infrared (62211)
          Group 53 Created with Sketch.
          Multispecies ERK1/ERK2 In-Cell ELISA Kit, Near Infrared (62211)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Multispecies ERK1/ERK2 In-Cell ELISA Kit, Near Infrared

          In-cell ELISA Kit {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Multispecies ERK1/ERK2 In-Cell ELISA Kit, Near Infrared (62211)
          videoThumbnail
          ►

          Product Specifications

          Sample type/volume

          Supernatant
          10,000 cells

          Hands-on time

          1 hr 20 min

          Time-to-result

          24 hr

          Homogenous (no wash)

          No

          Instrument

          Fluorescent Microplate Reader

          Product size

          96 Tests

          Contents

          Blocking Buffer
          20X Tris Buffered Saline
          Surfact-Amps 20
          Surfact-Amps X-100
          DyLight® 680 Goat Anti-Mouse IgG (H+L)
          DyLight 800 Goat Anti-Rabbit IgG (H+L)
          Thin Plate Seal Assembly
          Components included only in the target-specific kits: Antibody #1 and Antibody #2

          Shipping conditions

          Wet ice

          Storage

          2-8°C

          Protein name

          ERK1/2

          Protein family

          Other Proteins

          Species (tested)

          Human

          Detector antibody conjugate

          Unconjugated

          Label or dye

          DyLight™ 680 , DyLight™ 800

          About This Kit

          The ERK1/2 Multispecies In-Cell ELISA Near Infrared Detection Kits provide a simple and convenient method for quantifying intracellular proteins in whole cells.

          Principle of the method

          This kit enables simultaneous detection of post-translational-modified protein (PTM) and the corresponding unmodified protein, or two different targets within the same well. Simultaneously detecting targets within an ELISA well eliminates variability caused by differences in cell plating. The expression levels of the protein(s) are monitored using primary antibodies specific to the targets and corresponding species-specific near-infrared Thermo Scientific DyLight-Conjugated Secondary Antibodies. Relative protein activation or inactivation is determined as a ratio of modified protein levels to the corresponding unmodified protein measured using a modification-specific antibody and an antibody to unmodified protein, respectively.

          Rigorous validation

          Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

          Target information

          ERK1 and ERK2 are widely expressed and are involved in the regulation of meiosis, mitosis, and postmitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. When growth factors bind to the receptor tyrosine kinase, Ras interacts with Raf, the serine/threonine protein kinase and activates it as well. Once actived, Raf phosphorylates serine residue in 2 further kinases, MEK1/2, which in turn phosphorylates tyrosine/threonine in extracellular-signal regulated kinase (ERK) 1/2. Upon activation, the ERKs either phosphorylate a number of cytoplasmic targets or migrate to the nucleus, where they phosphorylate and activate a number of transcription factors such as c-Fos and Elk-1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Gene aliases : ERK, ERK-1, ERK-2, ERK1, ERK2, ERT1, ERT2, HS44KDAP, HUMKER1A, MAPK2, NS13, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, p44-ERK1, p44-MAPK, P44ERK1, P44MAPK, PRKM1, PRKM2, PRKM3

          Gene ID : (Human) 5594, (Human) 5595

          Gene symbol : MAPK1, MAPK3

          Protein Aliases : erk1 erk2, ERK1b, p42, p42 MAP Kinase, Erk1, cancer susceptibility protein, ERK1c, Erk2, extracellular signal-regulated kinase 1, extracellular signal-regulated kinase 2, extracellular signal-related kinase 1, insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK 2, microtubule-associated protein 2 kinase, mitogen-activated protein kinase 2, protein tyrosine kinase ERK2, unnamed protein product

          UniProt ID (Human) P28482, (Human) P27361

          Recommended Products

          IL-6 Human Matched Antibody Pair

          Because you are interested in IL-6 Human ELISA Kit.

          SIZE

          10 x 96 tests

          PRICE

          USD 400.00

          Cat # CHC1263

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-pj9bv:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0