Nalgene™ Biohazardous Waste Containers
Nalgene™ Biohazardous Waste Containers
Nalgene™ Biohazardous Waste Containers
Nalgene™ Biohazardous Waste Containers
Thermo Scientific™

Nalgene™ Biohazardous Waste Containers

Conduct secondary containment of biohazardous waste with these biohazardous waste containers, which feature convenient one-step sterilization and enable one-handed disposal of materials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) OuterHeight (Metric)
6370-001557 L34 cm74 cm
6370-00045.5 L22 cm28 cm
6370-000519 L28 cm43 cm
Catalog number 6370-0015
Price (USD)
391.07
Online Exclusive
461.04
Save 69.97 (15%)
Each
Capacity (Metric):
57 L
Diameter (Metric) Outer:
34 cm
Height (Metric):
74 cm
Price (USD)
391.07
Online Exclusive
461.04
Save 69.97 (15%)
Each

Conduct secondary containment of biohazardous waste with Thermo Scientific™ Nalgene™ Biohazardous Waste Containers, which feature convenient one-step sterilization—no need to remove the bag. A port opening in the cover allows for one-handed disposal of materials, limiting exposure to infectious waste. The containers hold standard sized autoclave bags (not included).

  • Polypropylene copolymer (PPCO) components will not rust, dent or puncture like other materials
  • Complies with U.S. OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
  • Autoclavable/Biohazard
  • Symbol printed in English and Spanish

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

Specifications
Capacity (English)15 gal.
Certifications/ComplianceU.S. OSHA Standard 29 CFR Part 1910.1030
Diameter (English) Outer13.25 in.
Height (English)29 in.
Height (Metric)74 cm
No. per Case1
No. per Pack1
Capacity (Metric)57 L
Diameter (Metric) Outer34 cm
MaterialPolypropylene
SterilityNon-sterile
TypeWaste Container Biohazardous
Unit SizeEach