GLD Pro Gas Leak Detector
GLD Pro Gas Leak Detector
GLD Pro Gas Leak Detector
GLD Pro Gas Leak Detector
Thermo Scientific™
GLD Pro Gas Leak Detector
Quickly locate and identify gas leaks in your GC instrument with the Thermo Scientific™ GLD Pro Gas Leak Detector.
Catalog NumberFor Use With (Equipment)
66002-004Gas Chromatography Systems
Catalog number 66002-004
Price (JPY)
-
For Use With (Equipment):
Gas Chromatography Systems
Detect and identify gas leaks quickly with the Thermo Scientific™ GLD Pro Gas Leak Detector. Specifically designed for use with GC systems, it detects minute leaks of a wide range of laboratory gases. This hand-held, easy-to-use detector features both visual and audible alarms and utilizes an LED light display to indicate the intensity of the leak. The small probe detector is designed to reach difficult and confined locations.
Additional Product Information
  • Pushbutton on/off switch
  • Pushbutton zero function
  • Automatic shutoff (five minutes)
  • Rechargable battery (up to six hours operation)
  • Durable storage case
  • Probe holder
  • One-year warranty
Specifications
DescriptionGLD Pro Gas Leak Detector
For Use With (Application)GC
For Use With (Equipment)Gas Chromatography Systems
Product TypeGas Leak Detector
Unit SizeEach