SST Sample Loops for Split-Loop Autosamplers
Actual product may vary
Thermo Scientific™
SST Sample Loops for Split-Loop Autosamplers
Order the Thermo Scientific™ SST Sample Loop you need for use in Thermo Scientific™ Dionex™ UltiMate™ split-loop autosamplers. Available in sizes from 25 to 1000μL.
Catalog NumberSample Capacity
6820.24291000 μL
6820.2451100 μL
6820.245225 μL
6820.2453250 μL
6820.2454500 μL
Catalog number 6820.2429
Price (JPY)
-
Sample Capacity:
1000 μL
Order the Thermo Scientific™ SST Sample Loop you need for use in Thermo Scientific™ Dionex™ UltiMate™ split-loop autosamplers. Available in sizes from 25 to 1000μL, the stainless steel (SST) sample loops incorporate Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.
Specifications
For Use With (Equipment)Dionex UltiMate 3000 Split-Loop Autosampler WPS-3000(T)SL Semiprep
MaterialStainless Steel
Sample Capacity1000 μL
TypeSample Loop
Unit SizeEach