Product Image
Thermo Scientific™

Buffer Tubing for UltiMate™ 3000 Autosamplers, Biocompatible

Select this biocompatible version of buffer tubing in volumes from 50 to 2000μL for your biocompatible Thermo Scientific™ UltiMate™ 3000 Autosampler.
Catalog NumberSample Capacity
6821.00212000 μL
6821.00221000 μL
6821.001950 μL
6821.0020500 μL
Catalog number 6821.0021
Price (JPY)
-
Sample Capacity:
2000 μL

Choose Thermo Scientific™ Dionex™ Biocompatible Buffer Tubing for Thermo Scientific™ UltiMate™ 3000 Autosamplers, available in volumes from 50 to 2000μL. Select the biocompatible buffer tubing volume of choice your biocompatible LC system.

Specifications
Biocompatible VersionYes
For Use With (Equipment)Dionex UltiMate 3000 Autosamplers WPS-3000 TBPL Nano/Cap
Sample Capacity2000 μL
TypeBuffer Tubing
Unit SizeEach