Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: ITGHSYVVYG PLCNGATSVL FESTPVYPNA GRYWETVERL KINQFYGAPT AVRLLLKYGD AWVKKYDRSS LRTLGSVGEP INCEAWEWLH RVVGDSRCTL VDTWWQTETG GICIAPRPSE EGAEILPAMA MRPFFGIVPV LMDEKGSVME GSNVSGALCI SQAWPGMART IYGDHQRFVD AYFKAYPGYY FTGDGAYRTE GGYYQITGRM DDVINISGHR LGTAEIEDAI ADHPAVPESA VIGYPHDIKG EAAFAFIVVK DSAGDSDVVV QELKSMVATK IAKYAVPDEI LVVKRLPKTR SGKVMRRLLR KIITSEAQEL GDTTTLEDPS IIAEILSVYQ KCKDKQAAAK
This gene encodes a mitochondrial acetyl-CoA synthetase enzyme. A similar protein in mice plays an important role in the tricarboxylic acid cycle by catalyzing the conversion of acetate to acetyl CoA. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AceCS2; Acetate--CoA ligase 2; Acetyl-CoA synthetase 2; Acetyl-coenzyme A synthetase 2-like, mitochondrial; Acyl-CoA synthetase short-chain family member 1; Propionate--CoA ligase; unnamed protein product
Gene Aliases: 1110032O15Rik; Acas2; ACAS2L; ACECS1; AceCS2; AceCS2L; ACSS1; AI788978; KIAA1846
UniProt ID: (Human) Q9NUB1, (Mouse) Q99NB1
Entrez Gene ID: (Human) 84532, (Mouse) 68738, (Rat) 296259
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support