CD11c Monoclonal Antibody (N418), eFluor™ 506, eBioscience™ - FAQs
CD11c Monoclonal Antibody (N418), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD11c Monoclonal Antibody (N418), eFluor™ 506, eBioscience™ - FAQs (69-0114-80, 69-0114-82)