Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 506, eBioscience™ - FAQs
View additional product information for Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 506, eBioscience™ - FAQs (69-4321-82, 69-4321-80)