NanoDrop Ultra Installation & Service Options
Thermo Scientific™
NanoDrop Ultra Installation & Service Options
Rely on our trained experts to perform on-site installation, recertification, or preventative maintenance of your Thermo Scientific™ NanoDrop™ Ultra Spectrophotometers.
Catalog NumberIncludes
701-062593NanoDrop Ultra Digital Signature SW OQ
701-062591NanoDrop Ultra/UltraC – SciVault 2 + IQ/OQ install
701-062592NanoDrop UltraFL/UltraCFL – SciVault 2 + IQ/OQ install
701-062597ND Ultra Accalro Pro, Install On Site
701-062598ND Ultra Acclaro Pro, Install Upgrade Depot
Catalog number 701-062593
Price (JPY)
-
Includes:
NanoDrop Ultra Digital Signature SW OQ
Rely on our trained experts to perform on-site installation, recertification, or preventative maintenance of your Thermo Scientific™ NanoDrop™ Ultra Spectrophotometers. Services include on-site installation of Thermo Scientific™ NanoDrop™ Ultra and 21 CFR Part 11 compliance software, instrument installation qualification and operational qualification, and preventative maintenance.
All engineer labor and travel charges for installation within continental US are included. Additional charges may apply for outside of continental US or remote Canadian regions. Separate purchase of 21 CFR Part 11 compliance software for NanoDrop Ultra Spectrophotometers and NanoDrop Ultra IQ/OQ Kit required.
NanoDrop Ultra Qualified Installation
Features:
  • On-site installation of instrument
  • On-site installation of NanoDrop Ultra software
  • (Optional) On-site installation of Thermo Scientific™ SciVault™ 2 Software
  • (Optional) On-site installation of NanoDrop Ultra PC Software
  • Instrument installation Qualification and Operational Qualification
Requirements:
  • Separate purchase of SciVault 2 software for NanoDrop Ultra
  • Separately purchased model-specific NanoDrop Ultra IQ/OQ Kit
Exclusions:
  • Consumables such as cuvettes, pipettes, flash drives, printer paper
NanoDrop Ultra Digital Signature SW OQ
Features:
  • Execution of NanoDrop Ultra SciVault 2 OQ
Requirements:
  • Separate purchase of SciVault 2 software for NanoDrop Ultra
  • Purchase of NanoDrop Ultra Qualified Installation
NanoDrop Ultra OQ Recertification
Features:
  • Annual Operational Qualification
  • All engineer labor and travel charges for OQ within continental US. Additional charges may apply for outside of continental US or remote Canadian regions.
Requirements:
  • Separate purchase of SciVault 2 for NanoDrop Ultra (If not already purchased)
  • Separately purchased NanoDrop Ultra IQ/OQ Kit (if not already purchased)
  • If not purchasing a new IQ/OQ Kit, applicable diagnostic reagents and filter set must be purchased individually if not already owned.
    • All models: CHEM-PV-1 and CHEM-PR1-KIT.
    • Cuvette Models: 840-257300
Exclusions:
  • Consumables such as cuvettes, pipettes, flash drives, printer paper
ND Ultra Acclaro Pro Upgrade
Features:
  • Software add-on to extend the performance of the NanoDrop Ultra by allowing increased accuracy of high concentration measurements
  • Photometric Accuracy: 5% up to 550 A
  • Acclaro Pro on-site or factory calibration
Specifications
For Use With (Equipment)NanoDrop Ultra Spectrophotometers and Fluorometers
IncludesNanoDrop Ultra Digital Signature SW OQ
Unit SizeEach