CaptureSelect™ Biotin Anti-C1-Inhibitor Conjugate
Thermo Scientific™
CaptureSelect™ Biotin Anti-C1-Inhibitor Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-C1-Inhibitor Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human C1 esterase inhibitor (C1-Inh).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7103402500500 μg
7103402100100 μg
Catalog number 7103402500
Price (USD)
2,240.00
Each
Quantity:
500 μg
Price (USD)
2,240.00
Each
CaptureSelect Biotin Anti-C1-Inhibitor Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human C1 esterase inhibitor (C1-Inh). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize native and recombinant human C1 esterase inhibitor.
Applications for CaptureSelect Biotin Anti-C1-Inhibitor Conjugate include ELISA, immunoprecipitation with streptavidin-coupled DynaBeads, Gyrolab™-based immunoassays, and label-free detection using platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Ligand TypeAntibody
Product LineCaptureSelect
Quantity500 μg
Label TypeBiotin and Analogs
Target SpeciesHuman
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).