BioBasic™ 18 HPLC Columns
Thermo Scientific™

BioBasic™ 18 HPLC Columns

Use this 300Å wide-pore silica for the separation of small to medium proteins and other biomolecules.
Catalog NumberLength (Metric)Diameter (Metric)
72105-254630250 mm4.6 mm
72105-252130250 mm2.1 mm
72105-259270A250 mm20 mm
72105-151030150 mm1 mm
72105-05103050 mm1 mm
72105-152130150 mm2.1 mm
72105-102130100 mm2.1 mm
72105-05213050 mm2.1 mm
72105-154630150 mm4.6 mm
72105-101030100 mm1 mm
72105-01400110 mm4 mm
72105-01210110 mm2.1 mm
72105-01100110 mm1 mm
Catalog number 72105-254630
Price (JPY)
122,500
Each
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm

Separate small to medium peptides with Thermo Scientific™ BioBasic™ 18 LC Columns.

  • 300Å pore size for maximum performance with biomolecules
  • High peak capacity stationary phase
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeReversed Phase
EndcappedYes
Max. Pressure5800 psi (400 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Product TypeAnalytical Column
Stationary PhaseC18
Surface Area100 m²/g
Temperature60°C
USP TypeL1
Carbon Load9%
Diameter (Metric)4.6 mm
Length (Metric)250 mm
MaterialStainless Steel
Particle Size5 μm
PhaseReversed Phase
Pore Size300 Å
Product LineBioBasic
TechniqueBio-LC
pH2 to 9
Unit SizeEach