Nalgene™  LDPE, Round Carboy with Handle
Thermo Scientific™

Nalgene™ LDPE, Round Carboy with Handle

For Use With: Store and transport reagents; Closure Material: Polypropylene; Color: White
Catalog NumberQuantity
8210-0020PK
also known as 8210-0020
Case of 6
Catalog number 8210-0020PK
also known as 8210-0020
Price (JPY)
11,800
Pack of 1
Quantity:
Case of 6

Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.

  • Molded-in graduations of 5L and 1 gal. increments
  • Leakproof†

Includes:

White polypropylene screw cap and 83B closure

Ordering Information:

8210 Series manufactured in our state-of-the-art Suzhou, China facility and available for purchase in Asia.  All other products manufactured in Rochester, NY.


 

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (English)2.6 gal.
Closure MaterialPolypropylene
Closure Size83B
DescriptionRound LDPE Carboy w/handle (Manufactured in our state-of-the-art Suzhou, China facility
Diameter (Metric) Inner Neck64 mm
Diameter (Metric) Outer250 mm
For Use With (Application)Store and transport reagents
Height (Metric) With Closure389 mm
IncludesHandles
No. per Case6/Cs.
No. per Pack1 Ea.
QuantityCase of 6
AutoclavableNot Autoclavable
Capacity (Metric)10 L
ColorWhite
MaterialLDPE
Product TypeBiotainer
Unit SizePack of 1