Product Image
Invitrogen™

eBioscience™ Human Regulatory T Cell Staining Kit #3

This Human Regulatory T cell Staining Kit contains all of the buffers and monoclonal antibodies for CD4, CD25, and Foxp3Read more
Catalog NumberQuantity
88-8995-401 kit
Catalog number 88-8995-40
Price (JPY)
77,500
Online offer
Ends: 26-Jun-2026
129,200
Save 51,700 (40%)
Each
Quantity:
1 kit
This Human Regulatory T cell Staining Kit contains all of the buffers and monoclonal antibodies for CD4, CD25, and Foxp3 necessary to successfully stain and identify regulatory T cells from human peripheral blood cells.

The RPA-T4 monoclonal antibody reacts with human CD4, a 59 kDa glycoprotein found on the surface of the majority of thymocytes, a subset of mature T cells (T helper cells), and at lower levels on monocytes. The BC96 monoclonal antibody reacts with human CD25 (also known as interleukin-2 receptor alpha, IL-2R alpha), a 55 kDa surface protein expressed by early progenitors of T cells and B cells, by mature, activated T cells and B cells, and at constitutively high levels on regulatory T cells. The PCH101 monoclonal antibody reacts with the amino terminus of human Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2. Foxp3 is a 49-55 kDa protein and a member of the forkhead/winged-helix family of transcription factors. It was identified as the gene responsible for the X-linked lymphoproliferative disease observed in scurfy (sf) mice and in the human disorder, X-linked autoimmunity-allergic dysregulation syndrome (XLAAD). Constitutive expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

Reactivity/Species
Human

Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
For Use With (Application)Intracellular Staining, Flow Cytometry
Product LineeBioscience
Product TypeCell Staining Kit
Unit SizeEach
Contents & Storage
2° to 8°C. Light sensitive material. Use within 6 months of opening or by date indicated on the bottle.