Product Image
Invitrogen™
MagniSort™ Human CD8 Positive Selection Kit
The MagniSort™ Human CD8 Positive Selection Kit is designed for the magnetic separation of CD8+ cells by positive selection. ItRead more
Have Questions?
Catalog NumberQuantity
8802-6832-74200 tests
Catalog number 8802-6832-74
Price (HKD)
6,796.00
Online Exclusive
Ends: 30-Jun-2026
8,301.00
Save 1,505.00 (18%)
Each
Quantity:
200 tests
Price (HKD)
6,796.00
Online Exclusive
Ends: 30-Jun-2026
8,301.00
Save 1,505.00 (18%)
Each
The MagniSort™ Human CD8 Positive Selection Kit is designed for the magnetic separation of CD8+ cells by positive selection. It has been optimized for the isolation of CD8+ cells from human peripheral blood monocuclear cells, utilizing a biotinylated Anti-Human CD8 antibody and streptavidin-coated magnetic beads. CD8+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD8+ cells by decanting.
After positive selection, the purity of selected cells can be verified by staining with Anti-Human CD8, clones RPA-T8 or SK1.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD8+
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
Product TypePositive Selection Kit
Unit SizeEach