Product Image
Invitrogen™
MagniSort™ Mouse B cell Enrichment Kit
The MagniSort™ Mouse B cell Enrichment Kit is designed for the magnetic separation of mouse B cells by negative selection.Read more
Catalog NumberQuantity
8804-6827-74200 tests
Catalog number 8804-6827-74
Price (JPY)
96,400
Online offer
Ends: 25-Sep-2026
160,700
Save 64,300 (40%)
Each
Quantity:
200 tests
The MagniSort™ Mouse B cell Enrichment Kit is designed for the magnetic separation of mouse B cells by negative selection. It has been optimized for the isolation of mouse B cells from spleens or lymph nodes utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave B cells untouched and free in solution.
The MagniSort™ Mouse B cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Mouse CD3 Biotin
Anti-Mouse CD4 Biotin
Anti-Mouse CD8 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD49b Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse TER-119 Biotin
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeB Cells
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypeCell Enrichment Kit
Unit SizeEach