Pierce™ Mouse Embryonic Fibroblast Isolation Kit
Thermo Scientific™

Pierce™ Mouse Embryonic Fibroblast Isolation Kit

The Thermo Scientific Pierce Mouse Embryonic Fibroblast Isolation Kit provides isolation and culturing reagents for the optimal yield and viabilityRead more
Have Questions?
Catalog NumberQuantity
882791 L kit
Catalog number 88279
Price (USD)
502.00
Each
Quantity:
1 L kit
Request bulk or custom format
Price (USD)
502.00
Each
The Thermo Scientific Pierce Mouse Embryonic Fibroblast Isolation Kit provides isolation and culturing reagents for the optimal yield and viability of fibroblasts from mouse embryos.

Features of the Mouse Embryonic Fibroblast Isolation Kit:

• Optimized – procedure and reagents optimized for viability, yield, purity, and ease-of-use
• Yield – provides a 2-fold increase in yield compared to do-it-yourself methods and other kits
• Functional – cultured fibroblasts express the appropriate biochemical markers

Includes:
• Complete kit includes Fibroblast Isolation Enzyme, Hanks' Balanced Salt Solution (HBSS), and DMEM for Primary Cell Isolation

The Mouse Embryonic Fibroblast Isolation Kit has been designed to obtain high viability and purity of embryonic fibroblasts using validated reagents and an optimized procedure. The kit contains a tissue-specific dissociation enzyme and media formulated for primary cell culture that promotes the isolation growth of functional fibroblasts. In addition, MEF isolated and cultured with the Pierce MEF Isolation Kit can be cryopreserved for future use with very minimal loss of viability.

Applications:
• Generation of immortalized cell lines from transgenic mice
• Feeder layer for stem cells
• IHC and IF studies

Because it uses an optimized tissue-specific dissociation enzyme, this kit isolates fibroblasts with greater yield and cell viability than cells isolated by existing methods. Isolated MEF express fibroblast-specific protein markers, confirming their viability and unperturbed metabolic function. After isolation, the mouse fibroblasts may be immortalized by continuous passage for the establishment of unique cell lines. They may also be used as a cell culture model for a diverse range of studies such as gene regulation and stem cell research [1,2,3,4].

More Product Data
• An improved method for highly efficient isolation of primary mouse embryonic fibroblasts

Related Products
Mouse Embryonic Fibroblast Isolation Enzyme 1 (with papain) for Pierce™ Primary Cell Isolation Kits
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeFibroblasts
For Use With (Application)Stem dells, immunohistochemistry and immunofluorescence
Quantity1 L kit
Sufficient For50 Samples
Target SpeciesMouse
Product LinePierce
Product TypeCell Isolation Kit
Unit SizeEach
Contents & Storage
Sufficient For: Isolation of MEFs from 50 embryonic mouse somatic tissues
• DMEM for Primary Cell Isolation, 500 mL
• MEF Isolation Enzyme (with papain), 5 vials
• Hanks' Balanced Salt Solution (HBSS without Ca2+, Mg2+), 500 mL

Store kit at 4°C.

Frequently asked questions (FAQs)

What kits are available for cell isolation?

We offer the following kits for cell isolation:

Primary Cardiomyocyte Isolation Kit, Cat. No. 88281
Mouse Embryonic Fibroblast Isolation Kit, Cat. No. 88279
Primary Neuron Isolation Kit, Cat. No. 88280

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.