Mouse Embryonic Fibroblast Isolation Enzyme 1 (with papain) for Pierce™ Primary Cell Isolation Kits
Thermo Scientific™
Mouse Embryonic Fibroblast Isolation Enzyme 1 (with papain) for Pierce™ Primary Cell Isolation Kits
Thermo Scientific Pierce Mouse Embryonic Fibroblast Isolation Enzyme (with papain) is a tissue-specific dissociation reagent for isolation of MEFs fromRead more
Have Questions?
Catalog NumberQuantity
882901 Vial
Catalog number 88290
Price (USD)
87.25
Each
Quantity:
1 Vial
Request bulk or custom format
Price (USD)
87.25
Each
Thermo Scientific Pierce Mouse Embryonic Fibroblast Isolation Enzyme (with papain) is a tissue-specific dissociation reagent for isolation of MEFs from mouse embryonic tissue and is a component of the Thermo Scientific™ Pierce™ Mouse Embryonic Fibroblast Isolation Kit.
Papain has been demonstrated to be more effective and less harsh than other proteases for tissue dissociation. At its optimal concentration, this reagent ensures a high yield of viable and functional fibroblasts.
More Product Data
• An improved method for highly efficient isolation of primary mouse embryonic fibroblasts
Related Products
Pierce™ Mouse Embryonic Fibroblast Isolation Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeFibroblasts
Quantity1 Vial
Target SpeciesMouse
FormSolid
Product LinePierce
Product TypeCell Isolation Kit
Unit SizeEach
Contents & Storage
Upon receipt store product at 2-80°C. Product is shipped on ice packs.