Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Thermo Scientific™
Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems through the superior tip straightness and low %CV.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredTip StyleFit for PurposeUnit Size
912-262PK
also known as 912-262
10 μLNon-filteredStandardCertified QualityPack of 960
912-0218 μLFilteredStandardSterile and PCR ReadyCase of 4800
912-26110 μLNon-filteredStandardSterileCase of 4800
912-261PK
also known as 912-261
10 μLNon-filteredStandardSterilePack of 960
912-26210 μLNon-filteredStandardCertified QualityCase of 4800
916-02140 μLFilteredStandardSterile and PCR ReadyCase of 4800
916-021PK
also known as 916-021
40 μLFilteredStandardSterile and PCR ReadyPack of 960
916-26150 μLNon-filteredStandardSterileCase of 4800
916-261PK
also known as 916-261
50 μLNon-filteredStandardSterilePack of 960
916-26250 μLNon-filteredStandardCertified QualityCase of 4800
916-262PK
also known as 916-262
50 μLNon-filteredStandardCertified QualityPack of 960
916-262-0550 μLNon-filteredLow RetentionCertified QualityCase of 4800
916-262-05PK
also known as 916-262-05
50 μLNon-filteredLow RetentionCertified QualityPack of 960
918-02120 μLFilteredStandardSterile and PCR ReadyCase of 4800
918-021PK
also known as 918-021
20 μLFilteredStandardSterile and PCR ReadyPack of 960
918-021-0520 μLFilteredLow RetentionSterile and PCR ReadyCase of 4800
918-021-05PK
also known as 918-021-05
20 μLFilteredLow RetentionSterile and PCR ReadyPack of 960
918-26120 μLNon-filteredStandardSterileCase of 4800
918-261PK
also known as 918-261
20 μLNon-filteredStandardSterilePack of 960
918-261-0520 μLNon-filteredLow RetentionSterileCase of 4800
918-26220 μLNon-filteredStandardCertified QualityCase of 4800
918-262PK
also known as 918-262
20 μLNon-filteredStandardCertified QualityPack of 960
918-262-0520 μLNon-filteredLow RetentionCertified QualityCase of 4800
918-262-05PK
also known as 918-262-05
20 μLNon-filteredLow RetentionCertified QualityPack of 960
919-021130 μLFilteredStandardSterile and PCR ReadyCase of 4800
919-021PK
also known as 919-021
130 μLFilteredStandardSterile and PCR ReadyPack of 960
919-021-05130 μLFilteredLow RetentionSterile and PCR ReadyCase of 4800
919-021-05PK
also known as 919-021-05
130 μLFilteredLow RetentionSterile and PCR ReadyPack of 960
919-021G130 μLFilteredWide BoreSterile and PCR ReadyCase of 4800
919-261250 μLNon-filteredStandardSterileCase of 4800
919-261PK
also known as 919-261
250 μLNon-filteredStandardSterilePack of 960
919-261-05250 μLNon-filteredLow RetentionSterileCase of 4800
919-261-05PK
also known as 919-261-05
250 μLNon-filteredLow RetentionSterilePack of 960
919-261G250 μLNon-filteredWide BoreSterileCase of 4800
919-261GPK
also known as 919-261G
250 μLNon-filteredWide BoreSterilePack of 960
919-262250 μLNon-filteredStandardCertified QualityCase of 4800
919-262PK
also known as 919-262
250 μLNon-filteredStandardCertified QualityPack of 960
919-262-05250 μLNon-filteredLow RetentionCertified QualityCase of 4800
919-262-05PK
also known as 919-262-05
250 μLNon-filteredLow RetentionCertified QualityPack of 960
919-262G250 μLNon-filteredWide BoreCertified QualityCase of 4800
919-262GPK
also known as 919-262G
250 μLNon-filteredWide BoreCertified QualityPack of 960
276050 μLFilteredExtended LengthSterileCase of 4800
Catalog number 912-262PK
also known as 912-262
Price (USD)
143.37
Online Exclusive
159.30
Save 15.93 (10%)
Pack of 960
Volume (Metric):
10 μL
Filtered:
Non-filtered
Tip Style:
Standard
Fit for Purpose:
Certified Quality
Unit Size:
Pack of 960
Price (USD)
143.37
Online Exclusive
159.30
Save 15.93 (10%)
Pack of 960
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
Specifications
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
Packaging96 tips/rack, 10 racks/pack, 5 packs/case
Quantity4800 tips
AutoclavableAutoclavable
ColorClear
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Beckman Coulter Liquid Handling Systems
FormatRack
Product LineBeckman
SterilityNon-sterile
Tip StyleStandard
Volume (Metric)10 μL
Unit SizePack of 960