Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
Thermo Scientific™
Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Catalog NumberVolume (Metric)FilteredSterilityFit for Purpose
915-261PK
also known as 915-261
20 μLNon-filteredSterileSterile
915-021PK
also known as 915-021
20 μLFilteredSterileSterile and PCR Ready
915-262PK
also known as 915-262
20 μLNon-filteredNon-sterileCertified Quality
917-021PK
also known as 917-021
130 μLFilteredSterileSterile and PCR Ready
917-261PK
also known as 917-261
200 μLNon-filteredSterileSterile
917-262PK
also known as 917-262
200 μLNon-filteredNon-sterileCertified Quality
Catalog number 915-261PK
also known as 915-261
Price (JPY)
11,100
Online offer
Ends: 26-Jun-2026
13,860
Save 2,760 (20%)
Pack of 960
Volume (Metric):
20 μL
Filtered:
Non-filtered
Sterility:
Sterile
Fit for Purpose:
Sterile
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
Specifications
ColorClear
FilteredNon-filtered
Fit for PurposeSterile
For Use With (Equipment)Beckman Coulter Liquid Handling Systems
FormatRack
Product LineBeckman
SterilitySterile
Tip StyleExtended Length
Volume (Metric)20 μL
Unit SizePack of 960