Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
Thermo Scientific™
Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Catalog NumberVolume (Metric)FilteredSterilityFit for Purpose
923-021PK
also known as 923-021
150 μLFilteredSterileSterile and PCR Ready
920-021PK
also known as 920-021
50 μLFilteredSterileSterile and PCR Ready
920-261PK
also known as 920-261
50 μLNon-filteredSterileSterile
920-262PK
also known as 920-262
50 μLNon-filteredNon-sterileCertified Quality
921-021PK
also known as 921-021
80 μLFilteredSterileSterile and PCR Ready
921-261PK
also known as 921-261
100 μLNon-filteredSterileSterile
921-262PK
also known as 921-262
100 μLNon-filteredNon-sterileCertified Quality
923-261PK
also known as 923-261
200 μLNon-filteredSterileSterile
923-262PK
also known as 923-262
200 μLNon-filteredNon-sterileCertified Quality
Catalog number 923-021PK
also known as 923-021
Price (JPY)
20,320
Pack of 960
Volume (Metric):
150 μL
Filtered:
Filtered
Sterility:
Sterile
Fit for Purpose:
Sterile and PCR Ready
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Warranty:A five (5)-year shelf life from the date of manufacture is recommended.
Specifications
ColorClear
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Zymark/Caliper Liquid Handling Workstations
FormatRack
Product LineCaliper Zymark
SterilitySterile
Tip StyleAutomation
Volume (Metric)150 μL
Unit SizePack of 960