Acetonitrile, Certified AR for Analysis, Fisher Chemical™
Acetonitrile, Certified AR for Analysis, Fisher Chemical™
Acetonitrile, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Acetonitrile, Certified AR for Analysis, Fisher Chemical™

C2H3N, CAS Number-75-05-8, 5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Metal can, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C
Quantity:
2.5 L
5 L
Packaging:
View all
Plastic-coated Glass Bottle
Catalog number A-0621-PB17
also known as A/0621/PB17
Price (JPY)
-
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Specifications
Boiling Point81°C
CAS75-05-8
ColorColorless
Formula Weight41.05g/mol
IUPAC Nameacetonitrile
View more
C2H3N, CAS Number-75-05-8, 5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Metal can, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C