Iso-Amyl Alcohol, Extra Pure, SLR, for Determination Of Fat In Milk, Fisher Chemical™
Iso-Amyl Alcohol, Extra Pure, SLR, for Determination Of Fat In Milk, Fisher Chemical™
Iso-Amyl Alcohol, Extra Pure, SLR, for Determination Of Fat In Milk, Fisher Chemical™
Fisher Chemical™

Iso-Amyl Alcohol, Extra Pure, SLR, for Determination Of Fat In Milk, Fisher Chemical™

Quantity:
500 mL
2.5 L
Packaging:
View all
Amber Glass Bottle
Catalog number A-7000-08TS
also known as A/7000/08
Price (JPY)
-
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
CAS123-51-3
Formula Weight88.15g/mol
MDL Number2934
Melting Point-117°C
Molecular FormulaC5H12O
View more
Iso-Amyl Alcohol, Extra Pure, SLR, for Determination Of Fat In Milk