L-Alanine methyl ester hydrochloride, 99%
L-Alanine methyl ester hydrochloride, 99%
L-Alanine methyl ester hydrochloride, 99%
Thermo Scientific Chemicals

L-Alanine methyl ester hydrochloride, 99%

CAS: 2491-20-5 | C4H10ClNO2 | 139.58 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A10178.14
also known as A10178-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2491-20-5
IUPAC Namemethyl (2S)-2-aminopropanoate hydrochloride
Molecular FormulaC4H10ClNO2
InChI KeyIYUKFAFDFHZKPI-DOIAFFBONA-N
SMILESCl.COC(=O)[C@H](C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥98.5%
Water Content (Karl Fischer Titration)≤1.5%
Optical Rotation+6° to +9° (c=1.6 in methanol)
View more
L-Alanine methyl ester hydrochloride is used to make in-vivo measurement of glucose and alanine metabolism in studies of patients with diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solubility
Soluble in Water (100 mg/ml).

Notes
Hygroscopic store under inert gas Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Hironobu Murata; Fumio Sanda; and Takeshi Endo. Highly Radical-Polymerizable Methacrylamide Having Dipeptide Structure. Synthesis and Radical Polymerization of N-Methacryloyl-l-leucyl-l-alanine Methyl Ester. Macromolecules. 1996, 29,(17), 5535-5538.
  2. Hongwu Zhao; and Wenting Hua. Synthesis and Characterization of Pyridine-Based Polyamido-Polyester Optically Active Macrocycles and Enantiomeric Recognition for d- andl-Amino Acid Methyl Ester Hydrochloride. J. Org. Chem. 2000, 65,(10), 2933-2938.